Search
Search

COPG1| Gene Symbol: COPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030597.2 | NP_001025768.1 | 15 - 16 | 1660 | 86 | |
| GenBank mRNA | AJ851498.1 | - | 15 - 16 | 1661 | 86 | |
Set Membership: |
|