Search
Search

ANP32B| Gene Symbol: ANP32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030934.1 | NP_001026105.1 | 2 - 3 | 326 | 64 | |
| XM_015299813.2 | XP_015155299.1 | 2 - 3 | 343 | 64 | ||
| GenBank mRNA | AJ719354.1 | - | 2 - 3 | 355 | 64 | |
| EF424417.2 | - | 2 - 3 | 64 | |||
Set Membership: |
|