Search
Search

ASXL2| Gene Symbol: ASXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031096.1 | NP_001026267.1 | 11 - 12 | 2029 | 105 | |
| XM_015285098.2 | XP_015140584.1 | 11 - 12 | 2037 | 105 | ||
| XM_015285099.2 | XP_015140585.1 | 13 - 14 | 1928 | 105 | ||
| XM_015285100.2 | XP_015140586.1 | 8 - 9 | 1622 | 105 | ||
| XM_025148790.1 | XP_025004558.1 | 11 - 12 | 1875 | 105 | ||
| XM_025148791.1 | XP_025004559.1 | 11 - 12 | 1754 | 105 | ||
| XM_025148792.1 | XP_025004560.1 | 10 - 11 | 1910 | 105 | ||
| GenBank mRNA | AJ719496.1 | - | 11 - 12 | 2029 | 105 | |
Set Membership: |
|