Search
Search

STK26| Gene Symbol: STK26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031117.1 | NP_001026288.1 | 1 - 2 | 78 | 78 | |
| XM_015278244.2 | XP_015133730.1 | 2 - 3 | 401 | 78 | ||
| GenBank mRNA | AJ719477.1 | - | 1 - 2 | 78 | 78 | |
Set Membership: |
|