Search
Search

INSIG2| Gene Symbol: INSIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305465.1 | NP_001292394.1 | 4 - 5 | 660 | 77 | |
| NM_001306186.1 | NP_001293115.1 | 4 - 5 | 500 | 77 | ||
| NM_001306187.1 | NP_001293116.1 | 3 - 4 | 384 | 77 | ||
| XM_015289825.2 | XP_015145311.1 | 4 - 5 | 693 | 77 | ||
| GenBank mRNA | AJ851538.1 | - | 4 - 5 | 77 | ||
| BX933359.1 | - | 4 - 5 | 77 | |||
Set Membership: |
|