Search
Search

FAM20B| Gene Symbol: FAM20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031268.1 | NP_001026439.1 | 3 - 4 | 690 | 56 | |
| XM_015290281.2 | XP_015145767.1 | 4 - 5 | 815 | 56 | ||
| GenBank mRNA | AJ720888.1 | - | 3 - 4 | 690 | 56 | |
Set Membership: |
|