Search
Search

TMEM65| Gene Symbol: TMEM65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031507.1 | NP_001026678.1 | 6 - 7 | 561 | 59 | |
| XM_015283110.2 | XP_015138596.1 | 8 - 9 | 1169 | 59 | ||
| XM_015283111.2 | XP_015138597.1 | 7 - 8 | 1098 | 59 | ||
| GenBank mRNA | AJ719948.1 | - | 6 - 7 | 561 | 59 | |
| CR391308.1 | - | 6 - 7 | 388 | 59 | ||
Set Membership: |
|