Search
Search

LOC431499
LOC100859910
LOC107050349
LOC107050798| Gene Symbol: LOC431499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031606.1 | NP_001026777.1 | 5 - 6 | 1439 | 91 | |
| GenBank mRNA | AJ851601.1 | - | 5 - 6 | 1439 | 91 | |
| Gene Symbol: LOC100859910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155943.1 | XP_025011711.1 | 5 - 6 | 1581 | 91 | |
| XM_025155944.1 | XP_025011712.1 | 5 - 6 | 1575 | 91 | ||
| Gene Symbol: LOC107050349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155945.1 | XP_025011713.1 | 5 - 6 | 1555 | 91 | |
| Gene Symbol: LOC107050798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155991.1 | XP_025011759.1 | 6 - 7 | 1076 | 91 | |
Set Membership: |
|