Search
Search

NFE2L1| Gene Symbol: NFE2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030756.1 | NP_001025927.1 | 6 - 7 | 1182 | 59 | |
| GenBank mRNA | AJ719867.1 | - | 6 - 7 | 1182 | 59 | |
| CR354056.1 | - | 4 - 5 | 916 | 59 | ||
Set Membership: |
|