Search
Search

INSIG1| Gene Symbol: INSIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030966.1 | NP_001026137.1 | 3 - 4 | 513 | 79 | |
| XM_025147161.1 | XP_025002929.1 | 2 - 3 | 731 | 79 | ||
| GenBank mRNA | AJ719295.1 | - | 3 - 4 | 513 | 79 | |
Set Membership: |
|