Search
Search

PPP1R2| Gene Symbol: PPP1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031313.1 | NP_001026484.1 | 4 - 5 | 473 | 114 | |
| XM_015291390.2 | XP_015146876.1 | 4 - 5 | 484 | 114 | ||
| GenBank mRNA | AJ851823.1 | - | 4 - 5 | 473 | 114 | |
Set Membership: |
|