Search
Search

STUB1| Gene Symbol: STUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031406.2 | NP_001026577.1 | 1 - 2 | 277 | 96 | |
| XM_015294667.1 | XP_015150153.1 | 1 - 2 | 272 | 96 | ||
| GenBank mRNA | AJ720999.1 | - | 1 - 2 | 277 | 96 | |
Set Membership: |
|