Search
Search

MORF4L2| Gene Symbol: MORF4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037173.1 | NP_001032250.1 | 3 - 4 | 181 | 105 | |
| NM_001037176.1 | NP_001032253.1 | 3 - 4 | 181 | 105 | ||
| GenBank mRNA | BI389851.1 | - | 3 - 4 | 173 | 105 | |
| BI391102.1 | - | 3 - 4 | 181 | 105 | ||
| BX930523.1 | - | 3 - 4 | 173 | 105 | ||
| CR386790.1 | - | 3 - 4 | 145 | 105 | ||
Set Membership: |
|