Search
Search

PRPF19| Gene Symbol: PRPF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039331.2 | NP_001034420.2 | 10 - 11 | 924 | 63 | |
| GenBank mRNA | AJ719482.1 | - | 10 - 11 | 927 | 63 | |
Set Membership: |
|