Search
Search

IL13| Gene Symbol: IL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007085.2 | NP_001007086.1 | 3 - 4 | 302 | 80 | |
| NM_001195791.1 | NP_001182720.1 | 2 - 3 | 251 | 80 | ||
| GenBank mRNA | AJ621250.1 | - | 3 - 4 | 302 | 80 | |
| GU119894.1 | - | 2 - 3 | 251 | 80 | ||
Set Membership: |
|