Search
Search

CDC2L1| Gene Symbol: CDC2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030871.3 | NP_001026042.2 | 10 - 11 | 1216 | 91 | |
| XM_015296863.2 | XP_015152349.1 | 11 - 12 | 4066 | 91 | ||
| XM_015296864.2 | XP_015152350.1 | 11 - 12 | 4063 | 91 | ||
| XM_015296865.2 | XP_015152351.1 | 11 - 12 | 4060 | 91 | ||
| XM_015296866.2 | XP_015152352.1 | 10 - 11 | 3943 | 91 | ||
| XM_025142506.1 | XP_024998274.1 | 11 - 12 | 4084 | 91 | ||
| XM_025142508.1 | XP_024998276.1 | 10 - 11 | 3940 | 91 | ||
Set Membership: |
|