Search
Search

GPR39| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080105.1 | NP_001073574.1 | 1 - 2 | 1222 | 65 | |
| XM_015289830.2 | XP_015145316.1 | 2 - 3 | 1173 | 65 | ||
| GenBank mRNA | DQ768750.1 | - | 1 - 2 | 1222 | 65 | |
Set Membership: |
|