Search
Search

ARR3| Gene Symbol: ARR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097533.1 | NP_001091002.1 | 11 - 12 | 872 | 80 | |
| XM_025149753.1 | XP_025005521.1 | 7 - 8 | 951 | 80 | ||
| GenBank mRNA | EF457884.1 | - | 11 - 12 | 872 | 80 | |
Set Membership: |
|