Search
Search

SPINK7| Gene Symbol: SPINK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112662.1 | NP_001106132.1 | 7 - 8 | 623 | 113 | |
| NM_001308494.1 | NP_001295423.1 | 7 - 8 | 629 | 113 | ||
| XM_015293763.2 | XP_015149249.1 | 8 - 9 | 768 | 113 | ||
| XM_015293764.2 | XP_015149250.1 | 8 - 9 | 767 | 113 | ||
| GenBank mRNA | BX260174.3 | - | 3 - 4 | 277 | 113 | |
| BX277920.4 | - | 7 - 8 | 623 | 113 | ||
| J00902.1 | - | 7 - 8 | 643 | 113 | ||
Set Membership: |
|