Search
Search

TCF12| Gene Symbol: TCF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205375.4 | NP_990706.2 | 15 - 16 | 1769 | 66 | |
| XM_015278764.2 | XP_015134250.2 | 20 - 21 | 2222 | 66 | ||
| XM_015278765.2 | XP_015134251.2 | 20 - 21 | 2222 | 66 | ||
| XM_015278766.2 | XP_015134252.2 | 20 - 21 | 2219 | 66 | ||
| XM_015278767.2 | XP_015134253.2 | 20 - 21 | 2219 | 66 | ||
| XM_015278768.2 | XP_015134254.2 | 19 - 20 | 2150 | 66 | ||
| XM_015278769.2 | XP_015134255.2 | 19 - 20 | 2147 | 66 | ||
| XM_015278770.2 | XP_015134256.2 | 19 - 20 | 2147 | 66 | ||
| XM_015278771.1 | XP_015134257.1 | 12 - 13 | 1572 | 66 | ||
| XM_015278772.2 | XP_015134258.1 | 11 - 12 | 1480 | 66 | ||
| XM_015278773.2 | XP_015134259.1 | 11 - 12 | 1477 | 66 | ||
| XM_025153805.1 | XP_025009573.1 | 20 - 21 | 2232 | 66 | ||
| XM_025153806.1 | XP_025009574.1 | 12 - 13 | 1471 | 66 | ||
| GenBank mRNA | M87337.1 | - | 11 - 12 | 1385 | 66 | |
| M97634.1 | - | 8 - 9 | 1003 | 66 | ||
| U12974.1 | - | 11 - 12 | 1377 | 66 | ||
| X64295.1 | - | 1385 | 66 | |||
Set Membership: |
|