Search
Search

GTF3A| Gene Symbol: GTF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113571.1 | NP_001107043.1 | 2 - 3 | 383 | 112 | |
| GenBank mRNA | CR352757.1 | - | 2 - 3 | 386 | 112 | |
| CR406086.1 | - | 1 - 2 | 69 | 112 | ||
Set Membership: |
|