Search
Search

HPSE| Gene Symbol: HPSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204167.1 | NP_989498.1 | 1 - 2 | 190 | 53 | |
| XM_025149456.1 | XP_025005224.1 | 1 - 2 | 202 | 53 | ||
| GenBank mRNA | AY037007.1 | - | 1 - 2 | 190 | 53 | |
Set Membership: |
|