Search
Search

SMN| Gene Symbol: SMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204199.1 | NP_989530.1 | 1 - 2 | 40 | 66 | |
| XM_015280581.2 | XP_015136067.1 | 1 - 2 | 62 | 66 | ||
| XM_015280582.2 | XP_015136068.1 | 1 - 2 | 62 | 66 | ||
| GenBank mRNA | AF322650.1 | - | 1 - 2 | 40 | 66 | |
Set Membership: |
|