Search
Search

ENTPD8| Gene Symbol: ENTPD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204247.2 | NP_989578.2 | 6 - 7 | 1108 | 127 | |
| XM_015279254.2 | XP_015134740.1 | 6 - 7 | 3094 | 127 | ||
| XM_015279258.2 | XP_015134744.1 | 6 - 7 | 3107 | 127 | ||
| XM_025156014.1 | XP_025011782.1 | 6 - 7 | 2903 | 127 | ||
| XM_025156015.1 | XP_025011783.1 | 6 - 7 | 2891 | 127 | ||
| XM_025156016.1 | XP_025011784.1 | 5 - 6 | 4060 | 127 | ||
| XM_025156017.1 | XP_025011785.1 | 6 - 7 | 1764 | 127 | ||
| XM_025156018.1 | XP_025011786.1 | 5 - 6 | 4114 | 127 | ||
| GenBank mRNA | AF041355.1 | - | 5 - 6 | 779 | 127 | |
| AF426405.1 | - | 5 - 6 | 779 | 127 | ||
Set Membership: |
|