Search
Search

FABP4| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204290.1 | NP_989621.1 | 2 - 3 | 283 | 78 | |
| GenBank mRNA | AF432506.1 | - | 2 - 3 | 283 | 78 | |
| KT165375.1 | - | 2 - 3 | 256 | 78 | ||
Set Membership: |
|