Search
Search

SOSTDC1| Gene Symbol: SOSTDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204373.1 | NP_989704.1 | 1 - 2 | 346 | 100 | |
| XM_015281261.2 | XP_015136747.1 | 1 - 2 | 392 | 100 | ||
| GenBank mRNA | AY319926.1 | - | 1 - 2 | 346 | 100 | |
| BX933507.1 | - | 1 - 2 | 322 | 100 | ||
Set Membership: |
|