Search
Search

FGFBP2| Gene Symbol: FGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204447.1 | NP_989778.1 | 1 - 2 | 642 | 79 | |
| XM_015285726.2 | XP_015141212.1 | 2 - 3 | 1638 | 79 | ||
| GenBank mRNA | AY164487.1 | - | 1 - 2 | 642 | 79 | |
| BX929579.1 | - | 1 - 2 | 1098 | 79 | ||
| BX930678.2 | - | 1 - 2 | 680 | 79 | ||
| BX930841.2 | - | 1 - 2 | 677 | 79 | ||
Set Membership: |
|