Search
Search

RHAG| Gene Symbol: RHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204464.1 | NP_989795.1 | 9 - 10 | 1267 | 142 | |
| XM_015285084.2 | XP_015140570.1 | 9 - 10 | 1289 | 142 | ||
| GenBank mRNA | AF531095.1 | - | 9 - 10 | 1267 | 142 | |
Set Membership: |
|