Search
Search

EIF2AK2| Gene Symbol: EIF2AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204487.1 | NP_989818.1 | 1 - 2 | 513 | 77 | |
| XM_015283610.2 | XP_015139096.1 | 2 - 3 | 569 | 77 | ||
| XM_015283611.2 | XP_015139097.1 | 2 - 3 | 646 | 77 | ||
| XM_015283612.2 | XP_015139098.1 | 2 - 3 | 569 | 77 | ||
| GenBank mRNA | AB125660.1 | - | 1 - 2 | 513 | 77 | |
Set Membership: |
|