Search
Search

MTF2| Gene Symbol: MTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204508.1 | NP_989839.1 | 7 - 8 | 758 | 70 | |
| XM_015290601.2 | XP_015146087.1 | 7 - 8 | 937 | 70 | ||
| XM_015290602.2 | XP_015146088.1 | 5 - 6 | 553 | 70 | ||
| XM_015290604.2 | XP_015146090.1 | 5 - 6 | 546 | 70 | ||
| GenBank mRNA | AJ720132.1 | - | 7 - 8 | 942 | 70 | |
| AY251284.1 | - | 7 - 8 | 758 | 70 | ||
| BX930939.2 | - | 5 - 6 | 471 | 70 | ||
Set Membership: |
|