Search
Search

TIMD4| Gene Symbol: TIMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006149.1 | NP_001006149.1 | 1 - 2 | 92 | 86 | |
| GenBank mRNA | AJ719444.1 | - | 1 - 2 | 92 | 86 | |
| HG425165.1 | - | 1 - 2 | 50 | 86 | ||
| LN831080.1 | - | 1 - 2 | 50 | 86 | ||
| LN831081.1 | - | 1 - 2 | 50 | 86 | ||
| LN831082.1 | - | 1 - 2 | 50 | 86 | ||
Set Membership: |
|