Search
Search

YWHAE| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006219.1 | NP_001006219.1 | 2 - 3 | 375 | 133 | |
| XM_015295739.2 | XP_015151225.1 | 2 - 3 | 364 | 133 | ||
| GenBank mRNA | AJ719304.1 | - | 2 - 3 | 375 | 133 | |
Set Membership: |
|