Search
Search

FGF14| Gene Symbol: FGF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204777.1 | NP_990108.1 | 4 - 5 | 650 | 77 | |
| XM_015274261.2 | XP_015129747.1 | 4 - 5 | 655 | 77 | ||
| XM_015274265.2 | XP_015129751.1 | 3 - 4 | 1088 | 77 | ||
| XM_015274273.2 | XP_015129759.1 | 4 - 5 | 751 | 77 | ||
| XM_025141760.1 | XP_024997528.1 | 5 - 6 | 790 | 77 | ||
| XM_025141796.1 | XP_024997564.1 | 5 - 6 | 542 | 77 | ||
| XM_025141824.1 | XP_024997592.1 | 4 - 5 | 503 | 77 | ||
| XM_025141858.1 | XP_024997626.1 | 4 - 5 | 743 | 77 | ||
| XM_025141879.1 | XP_024997647.1 | 5 - 6 | 658 | 77 | ||
| GenBank mRNA | AF199605.1 | - | 4 - 5 | 580 | 77 | |
| AF199606.1 | - | 4 - 5 | 650 | 77 | ||
Set Membership: |
|