Search
Search

SPIN1Z| Gene Symbol: SPIN1Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204633.1 | NP_989964.1 | 5 | 2552 | 147 | |
| XM_015280236.2 | XP_015135722.1 | 6 | 2918 | 147 | ||
| XM_015280237.2 | XP_015135723.1 | 6 | 3074 | 147 | ||
| XM_015280238.2 | XP_015135724.1 | 6 | 2733 | 147 | ||
| XM_015280239.2 | XP_015135725.1 | 6 | 2624 | 147 | ||
| XM_015280240.2 | XP_015135726.1 | 8 | 3022 | 147 | ||
| XM_025144640.1 | XP_025000408.1 | 6 | 2702 | 147 | ||
| XM_025144641.1 | XP_025000409.1 | 7 | 3221 | 147 | ||
| XM_025144642.1 | XP_025000410.1 | 7 | 3146 | 147 | ||
| XM_025144643.1 | XP_025000411.1 | 6 | 2999 | 147 | ||
| XM_025144644.1 | XP_025000412.1 | 5 | 2516 | 147 | ||
| GenBank mRNA | AB047853.1 | - | 5 | 2552 | 147 | |
Set Membership: |
|