Search
Search

QRFPR| Gene Symbol: QRFPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127170.1 | NP_001120642.1 | 4 - 5 | 803 | 80 | |
| GenBank mRNA | BX950747.1 | - | 1 - 2 | 800 | 80 | |
| EU525171.1 | - | 4 - 5 | 803 | 80 | ||
| KY856722.1 | - | 4 - 5 | 941 | 80 | ||
| KY856723.1 | - | 4 - 5 | 941 | 80 | ||
| KY856724.1 | - | 4 - 5 | 941 | 80 | ||
| KY856725.1 | - | 3 - 4 | 879 | 80 | ||
| KY856726.1 | - | 2 - 3 | 720 | 80 | ||
Set Membership: |
|