Search
Search

COL22A1| Gene Symbol: COL22A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139439.1 | NP_001132911.1 | 56 - 57 | 4174 | 58 | |
| XM_015283174.1 | XP_015138660.1 | 56 - 57 | 4298 | 58 | ||
| XM_025147123.1 | XP_025002891.1 | 55 - 56 | 4250 | 58 | ||
| XM_025147124.1 | XP_025002892.1 | 56 - 57 | 4298 | 58 | ||
Set Membership: |
|