Search
Search

FAM222B| Gene Symbol: FAM222B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030718.1 | NP_001025889.1 | 2 | 3050 | 69 | |
| XM_015295758.2 | XP_015151244.1 | 3 | 3195 | 69 | ||
| XM_015295759.2 | XP_015151245.1 | 3 | 3229 | 69 | ||
| XM_015295760.2 | XP_015151246.1 | 3 | 3479 | 69 | ||
| XM_015295761.2 | XP_015151247.1 | 3 | 3205 | 69 | ||
| XM_025141958.1 | XP_024997726.1 | 2 | 3104 | 69 | ||
| GenBank mRNA | AJ720625.1 | - | 2 | 3050 | 69 | |
Set Membership: |
|