Search
Search

GBX1| Gene Symbol: GBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205068.1 | NP_990399.1 | 2 | 1050 | 97 | |
| GenBank mRNA | AF022151.1 | - | 2 | 1050 | 97 | |
| HQ436511.1 | - | 1 | 417 | 97 | ||
Set Membership: |
|