Search
Search

TGFB1| Gene Symbol: TGFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318456.1 | NP_001305385.1 | 5 - 6 | 869 | 81 | |
| XM_025144453.1 | XP_025000221.1 | 6 - 7 | 1209 | 81 | ||
| GenBank mRNA | JQ423909.1 | - | 5 - 6 | 869 | 81 | |
Set Membership: |
|