Search
Search

MALRD1| Gene Symbol: MALRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318981.1 | NP_001305910.1 | 12 - 13 | 1566 | 77 | |
| GenBank mRNA | KC843480.1 | - | 12 - 13 | 1566 | 77 | |
Set Membership: |
|