Search
Search

PIP5K1C| Gene Symbol: PIP5K1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319021.1 | NP_001305950.1 | 15 - 16 | 1786 | 57 | |
| XM_015299762.2 | XP_015155248.1 | 15 - 16 | 1880 | 57 | ||
| XM_015299763.2 | XP_015155249.1 | 15 - 16 | 1883 | 57 | ||
| XM_015299764.2 | XP_015155250.1 | 15 - 16 | 1883 | 57 | ||
| XM_015299765.2 | XP_015155251.1 | 15 - 16 | 1880 | 57 | ||
| XM_015299766.2 | XP_015155252.1 | 15 - 16 | 1883 | 57 | ||
| GenBank mRNA | AB915768.1 | - | 15 - 16 | 1786 | 57 | |
| AB915769.1 | - | 15 - 16 | 1786 | 57 | ||
Set Membership: |
|