Search
Search

ITGA3| Gene Symbol: ITGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204146.1 | NP_989477.1 | 7 - 8 | 1137 | 78 | |
| XM_015299421.2 | XP_015154907.2 | 7 - 8 | 1221 | 78 | ||
| XM_015299422.2 | XP_015154908.2 | 7 - 8 | 1216 | 78 | ||
| XM_015299423.2 | XP_015154909.2 | 7 - 8 | 1221 | 78 | ||
| XM_015299424.2 | XP_015154910.2 | 7 - 8 | 1220 | 78 | ||
| XM_015299425.2 | XP_015154911.2 | 7 - 8 | 1219 | 78 | ||
| GenBank mRNA | AF330146.1 | - | 7 - 8 | 1137 | 78 | |
Set Membership: |
|