Search
Search

POR| Gene Symbol: POR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195796.1 | NP_001182725.1 | 15 - 16 | 2005 | 69 | |
| XM_015295712.2 | XP_015151198.1 | 16 - 17 | 2127 | 69 | ||
| XM_015295713.2 | XP_015151199.1 | 16 - 17 | 2149 | 69 | ||
| XM_015295714.2 | XP_015151200.1 | 16 - 17 | 2051 | 69 | ||
| XM_015295715.2 | XP_015151201.1 | 17 - 18 | 2991 | 69 | ||
| XM_015295717.2 | XP_015151203.1 | 17 - 18 | 2272 | 69 | ||
| XM_015295718.2 | XP_015151204.1 | 15 - 16 | 2053 | 69 | ||
| XM_015295719.2 | XP_015151205.1 | 14 - 15 | 2044 | 69 | ||
| XM_025141942.1 | XP_024997710.1 | 17 - 18 | 5130 | 69 | ||
| XM_025141943.1 | XP_024997711.1 | 19 - 20 | 5374 | 69 | ||
| XM_025141944.1 | XP_024997712.1 | 19 - 20 | 5388 | 69 | ||
| XM_025141945.1 | XP_024997713.1 | 18 - 19 | 5251 | 69 | ||
| XM_025141946.1 | XP_024997714.1 | 18 - 19 | 5253 | 69 | ||
| GenBank mRNA | HQ005378.1 | - | 14 - 15 | 1881 | 69 | |
Set Membership: |
|