Search
Search

SLCO2A1| Gene Symbol: SLCO2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198927.2 | NP_001185856.1 | 1 - 2 | 181 | 59 | |
| XM_025153357.1 | XP_025009125.1 | 5 - 6 | 1323 | 59 | ||
| XM_025153358.1 | XP_025009126.1 | 5 - 6 | 722 | 59 | ||
| GenBank mRNA | HQ380377.1 | - | 1 - 2 | 186 | 59 | |
Set Membership: |
|