Search
Search

MRPL21| Gene Symbol: MRPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199518.1 | NP_001186447.1 | 1 - 2 | 64 | 109 | |
| XM_015286208.2 | XP_015141694.1 | 1 - 2 | 75 | 109 | ||
| GenBank mRNA | BU247875.1 | - | 1 - 2 | 55 | 109 | |
Set Membership: |
|