Search
Search

RHOBTB1| Gene Symbol: RHOBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199608.1 | NP_001186537.1 | 6 - 7 | 1727 | 60 | |
| XM_015288129.2 | XP_015143615.1 | 8 - 9 | 1959 | 60 | ||
| XM_015288130.2 | XP_015143616.1 | 7 - 8 | 2657 | 60 | ||
| XM_025151547.1 | XP_025007315.1 | 8 - 9 | 2155 | 60 | ||
| XM_025151548.1 | XP_025007316.1 | 9 - 10 | 2002 | 60 | ||
| XM_025151549.1 | XP_025007317.1 | 7 - 8 | 1816 | 60 | ||
| XM_025151550.1 | XP_025007318.1 | 7 - 8 | 1940 | 60 | ||
Set Membership: |
|