Search
Search

QRICH1| Gene Symbol: QRICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257267.1 | NP_001244196.1 | 8 - 9 | 2120 | 73 | |
| XM_015292923.2 | XP_015148409.1 | 8 - 9 | 2177 | 73 | ||
| XM_015292924.2 | XP_015148410.1 | 8 - 9 | 2177 | 73 | ||
| XM_025154394.1 | XP_025010162.1 | 8 - 9 | 2434 | 73 | ||
| XM_025154395.1 | XP_025010163.1 | 9 - 10 | 2214 | 73 | ||
| XM_025154396.1 | XP_025010164.1 | 8 - 9 | 2158 | 73 | ||
| GenBank mRNA | CR354106.1 | - | 5 - 6 | 584 | 73 | |
| CR407006.1 | - | 4 - 5 | 442 | 73 | ||
Set Membership: |
|