Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg07160385_m1
          See other NRXN3 GE Assays ›
          Gene Symbol:
          NRXN3
          Gene Name:
          neurexin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 39728194 - 40668982 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg07160385_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NRXN3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271923.1 NP_001258852.1 17 - 18 3501 137
          NM_001271924.1 NP_001258853.1 2 - 3 540 137
          NM_001271925.1 NP_001258854.1 2 - 3 540 137
          XM_015287408.2 XP_015142894.2 19 - 20 4322 137
          XM_015287409.2 XP_015142895.2 19 - 20 4321 137
          XM_015287410.2 XP_015142896.2 18 - 19 4310 137
          XM_015287411.2 XP_015142897.2 19 - 20 4301 137
          XM_015287412.2 XP_015142898.2 18 - 19 4294 137
          XM_015287413.2 XP_015142899.2 18 - 19 4295 137
          XM_015287415.2 XP_015142901.2 18 - 19 4276 137
          XM_015287416.2 XP_015142902.2 19 - 20 4321 137
          XM_015287417.2 XP_015142903.2 16 - 17 4261 137
          XM_015287418.2 XP_015142904.2 18 - 19 4293 137
          XM_015287419.2 XP_015142905.2 17 - 18 4247 137
          XM_015287420.2 XP_015142906.2 16 - 17 4235 137
          XM_015287421.2 XP_015142907.2 19 - 20 4322 137
          XM_015287422.2 XP_015142908.2 16 - 17 4220 137
          XM_015287423.2 XP_015142909.2 18 - 19 4295 137
          XM_015287424.2 XP_015142910.2 18 - 19 4274 137
          XM_015287426.2 XP_015142912.2 16 - 17 4237 137
          XM_015287427.2 XP_015142913.2 19 - 20 4323 137
          XM_015287428.2 XP_015142914.2 19 - 20 4323 137
          XM_015287429.2 XP_015142915.2 19 - 20 4326 137
          XM_015287430.2 XP_015142916.2 19 - 20 4326 137
          XM_015287432.2 XP_015142918.1 19 - 20 4328 137
          XM_015287434.2 XP_015142920.1 19 - 20 4329 137
          XM_015287435.2 XP_015142921.1 19 - 20 4329 137
          XM_015287437.2 XP_015142923.2 16 - 17 4236 137
          XM_015287438.2 XP_015142924.2 18 - 19 4297 137
          XM_015287439.2 XP_015142925.1 16 - 17 4227 137
          XM_015287440.2 XP_015142926.2 18 - 19 4278 137
          XM_015287441.2 XP_015142927.2 18 - 19 4299 137
          XM_015287443.2 XP_015142929.2 2 - 3 1205 137
          XM_025150757.1 XP_025006525.1 18 - 19 4272 137
          XM_025150758.1 XP_025006526.1 17 - 18 4260 137
          XM_025150759.1 XP_025006527.1 17 - 18 4245 137
          XM_025150760.1 XP_025006528.1 14 - 15 4206 137
          XM_025150761.1 XP_025006529.1 16 - 17 4222 137
          XM_025150762.1 XP_025006530.1 15 - 16 4210 137
          XM_025150763.1 XP_025006531.1 14 - 15 4208 137
          XM_025150764.1 XP_025006532.1 13 - 14 4162 137
          XM_025150765.1 XP_025006533.1 17 - 18 4249 137
          XM_025150766.1 XP_025006534.1 17 - 18 4251 137
          XM_025150767.1 XP_025006535.1 18 - 19 4276 137
          XM_025150768.1 XP_025006536.1 14 - 15 4212 137
          XM_025150769.1 XP_025006537.1 13 - 14 4166 137
          XM_025150770.1 XP_025006538.1 18 - 19 4276 137
          XM_025150771.1 XP_025006539.1 17 - 18 4264 137
          XM_025150772.1 XP_025006540.1 17 - 18 4249 137
          XM_025150773.1 XP_025006541.1 17 - 18 4251 137
          XM_025150775.1 XP_025006543.1 16 - 17 4227 137
          XM_025150776.1 XP_025006544.1 19 - 20 4325 137
          GenBank mRNA BX932758.2 - 2 - 3 540 137
          EU702428.1 - 17 - 18 3501 137
          EU702429.1 - 2 - 3 426 137
          EU702430.1 - 2 - 3 540 137

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell adhesion
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline