Search
Search

SETD5| Gene Symbol: SETD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278017.2 | NP_001264946.2 | 13 - 14 | 1991 | 68 | |
| XM_015293099.2 | XP_015148585.1 | 14 - 15 | 2131 | 68 | ||
| XM_015293100.2 | XP_015148586.1 | 13 - 14 | 1856 | 68 | ||
| XM_025154480.1 | XP_025010248.1 | 12 - 13 | 1885 | 68 | ||
| GenBank mRNA | AJ733468.1 | - | 4 - 5 | 622 | 68 | |
| BU110740.1 | - | 3 - 4 | 421 | 68 | ||
Set Membership: |
|