Search
Search

PER3| Gene Symbol: PER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289779.2 | NP_001276708.1 | 8 - 9 | 1031 | 119 | |
| XM_004947315.3 | XP_004947372.1 | 8 - 9 | 981 | 119 | ||
| XM_004947316.3 | XP_004947373.1 | 8 - 9 | 981 | 119 | ||
| XM_004947317.3 | XP_004947374.1 | 8 - 9 | 981 | 119 | ||
| XM_004947319.3 | XP_004947376.1 | 8 - 9 | 981 | 119 | ||
| XM_015296850.2 | XP_015152336.1 | 9 - 10 | 982 | 119 | ||
| XM_015296851.2 | XP_015152337.1 | 8 - 9 | 882 | 119 | ||
| XM_025142495.1 | XP_024998263.1 | 8 - 9 | 981 | 119 | ||
| XM_025142496.1 | XP_024998264.1 | 8 - 9 | 981 | 119 | ||
| XM_417528.6 | XP_417528.4 | 8 - 9 | 981 | 119 | ||
Set Membership: |
|